Thermo Scientific Chemicals

L-Lysinamide dihydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H65741.03
1 g, Each
Thermo Scientific Chemicals

L-Lysinamide dihydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H65741.03
1 g, Each
Quantity
Catalog number: H65741.03
also known as H65741-03
Price (USD)
Price: 239.00
Your price:
Quantity
-

Chemical Identifiers

CAS
51127-08-3
IUPAC Name
(2S)-2,6-diaminohexanamide dihydrochloride
Molecular Formula
C6H17Cl2N3O
InChI Key
AIYVJLPYZQDCKV-XRIGFGBMSA-N
SMILES
Cl.Cl.NCCCC[C@H](N)C(N)=O
Assay from Suppliers CofA
≥94.0% (HPLC)
Form
Crystals or crystalline powder or powder
Appearance (Color)
White to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text