This product is animal origin and can only be shipped within the United States
Deoxyribonuclease I, bovine pancreas, Molecular Biology Grade
Thermo Scientific Chemicals

Deoxyribonuclease I, bovine pancreas, Molecular Biology Grade

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 U
500 U
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60027.EXR
also known as J60027-!R
Price (USD)
-
Quantity:
100 U
Request bulk or custom format
Chemical Identifiers
CAS9003-98-9
IUPAC NameDeoxyribonuclease
SMILES*
MDL NumberMFCD00130918
SpecificationsSpecification SheetSpecification Sheet
Unit definitionOne unit causes an increase in absorbance at 260nm of 0.001 per minute per ml, at 25?C and pH 5.0 when using highly polymerized DNA as a substrate
RNaseNone detected
Assay (unspecified)Units/mL: 2,000 min.
Protease activityNone detected
Endonuclease that splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide and removal of DNA from protein samples. Also used in is used to nick DNA, as a first step to incorporate labeled bases into DNA. The enzyme from Sigma has been used in the nuclease stock along with RNAse during cell lysate preparation of Madin-Darby canine kidney (MDCK) II cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only