Thermo Scientific Chemicals

L-Glutamine, Cell Culture Reagent

Catalog number: J60573.A1
1 kg, Each
Thermo Scientific Chemicals

L-Glutamine, Cell Culture Reagent

Catalog number: J60573.A1
1 kg, Each
Quantity
Catalog number: J60573.A1
also known as J60573-A1
Price (USD)
Price: 464.00
Online price: 393.65
Your price:
Quantity
-

Chemical Identifiers

CAS
56-85-9
IUPAC Name
(2S)-2-amino-4-carbamoylbutanoic acid
Molecular Formula
C5H10N2O3
InChI Key
ZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILES
N[C@@H](CCC(N)=O)C(O)=O
Assay (unspecified)
≥99.0%
Residue on ignition
≤0.1%
Impurity content
Total microorganisms: ≤100CFU/g
Appearance (Color)
White
Chloride content
≤0.02%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text