Thermo Scientific Chemicals

L-Histidine monohydrochloride monohydrate, Cell Culture Reagent, Thermo Scientific Chemicals

Catalog number: J61465.22
100 g, Each
Thermo Scientific Chemicals

L-Histidine monohydrochloride monohydrate, Cell Culture Reagent, Thermo Scientific Chemicals

Catalog number: J61465.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61465.22
also known as J61465-22
Price (USD)
Price: 109.00
Online price: 92.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5934-29-2
IUPAC Name
hydrogen (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrate chloride
Molecular Formula
C6H12ClN3O3
InChI Key
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
SMILES
[H+].O.[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O
Form
Powder or crystals or crystalline powder
Assay (special)
Chloride: 16.6-17.1%
Impurity content
Sulfate: ≤0.02%
pH
3.5-4.5
Assay from Supplier's CofA
≥98.0%

Description

L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

Solubility
Freely soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text