Thermo Scientific Chemicals

L-Phenylalanine, Cell Culture Reagent

Catalog number: J63925.30
250 g, Each
Thermo Scientific Chemicals

L-Phenylalanine, Cell Culture Reagent

Catalog number: J63925.30
250 g, Each
Quantity
Catalog number: J63925.30
also known as J63925-30
Price (USD)
Price: 223.00
Online price: 192.65
Your price:
Quantity
-

Chemical Identifiers

CAS
63-91-2
IUPAC Name
2-amino-3-phenylpropanoic acid
Molecular Formula
C9H11NO2
InChI Key
COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES
NC(CC1=CC=CC=C1)C(O)=O
Assay (special)
Sulfate: ≤0.02%
Form
Powder or crystals or crystalline powder
Assay (special)
Ammonium: ≤0.02%
Assay (special)
Endotoxin: ≤6.0 EU/g
Residue on ignition
≤0.1%

Description

The primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.

Solubility
Soluble in water. Aqueous solubility increases with warming. Also soluble in methanol, ethanol or 1M NH{4}OH

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text