Thermo Scientific Chemicals

Isomaltotriose, 97%

Catalog number: J64186.ME
500 mg, Each
Thermo Scientific Chemicals

Isomaltotriose, 97%

Catalog number: J64186.ME
500 mg, Each
Quantity
Catalog number: J64186.ME
also known as J64186-ME
Price (USD)
Price: 563.00
Your price:
Quantity
-

Chemical Identifiers

CAS
3371-50-4
IUPAC Name
(3R,4S,5S,6R)-6-({[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}methyl)oxane-2,3,4,5-tetrol
Molecular Formula
C18H32O16
InChI Key
FBJQEBRMDXPWNX-FYHZSNTMSA-N
SMILES
OC[C@H]1O[C@H](OC[C@H]2O[C@H](OC[C@H]3OC(O)[C@H](O)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Assay from Suppliers CofA
≥96.0%
Form
Powder or crystals or crystalline powder
Appearance (Color)
White to off-white

Description

Isomaltotriose is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isomaltotriose is used as pharmaceutical intermediate.

Solubility
Soluble in water

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text