Thermo Scientific Chemicals

Sucrose, ultrapure, 99%

Catalog number: J64270.A1
1 kg, Each
Thermo Scientific Chemicals

Sucrose, ultrapure, 99%

Catalog number: J64270.A1
1 kg, Each
Quantity
Catalog number: J64270.A1
also known as J64270-A1
Price (USD)
Price: 87.10
Online price: 74.65
Your price:
Quantity
-

Chemical Identifiers

CAS
57-50-1
IUPAC Name
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
CZMRCDWAGMRECN-PWPRYFECNA-N
SMILES
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay from Supplier's CofA
≥98.5%
Impurity content
Sulfate, SO4: ≤0.006% (≤60ppm)
Optical Rotation
+66.3 to +66.8°

Description

Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

Solubility
Soluble in water(1970g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text