Thermo Scientific Chemicals

N4-Benzoylcytidine, 99%

Catalog number: J65790.09
10 g, Each
Thermo Scientific Chemicals

N4-Benzoylcytidine, 99%

Catalog number: J65790.09
10 g, Each
Quantity
Catalog number: J65790.09
also known as J65790-09
Price (USD)
Price: 127.00
Online price: 110.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13089-48-0
Assay (HPLC)
98.5% min.
Water Content (Karl Fischer Titration)
5.0% max.
Loss on Drying
1.0% max.

Description

N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.

Solubility
Soluble in 1N NaOH at 10mg/ml. Also soluble in methanol or DMSO

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text