Thermo Scientific™
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide
Catalog number: J65878.MB
25 mg, Each
Thermo Scientific™
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide
Catalog number: J65878.MB
25 mg, Each
Quantity
Catalog number: J65878.MB
also known as J65878-MB
Price (USD)
Price: 334.00
Online price: 283.65
Your price:
Quantity
-
Description
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide is an chromogenic substrate for Caspase 3 and CCP-32.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.