Thermo Scientific Chemicals
C16 Ceramide, 98%
Catalog number: J67093.MCR
1 mg, Each
Thermo Scientific Chemicals
C16 Ceramide, 98%
Catalog number: J67093.MCR
1 mg, Each
Quantity
Catalog number: J67093.MCR
also known as J67093-M^
Price (USD)
Description
An endogenous ceramide that acts as a lipid second messenger to regulate apoptosis and stress signaling.