Krebs-Ringer Solution, bicarbonate-buffered
Thermo Scientific Chemicals
Krebs-Ringer Solution, bicarbonate-buffered
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J67591.K2
also known as J67591-K2
Price (USD)
164.65
Online exclusive
183.00
Save 18.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
164.65
Online exclusive
183.00
Save 18.35
Each
Specifications
PackagingPlastic Bottle
Recommended StorageAmbient Temperatures
Physical FormLiquid
Quantity1 L
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What is the pH of the Krebs-Ringer Solution, bicarbonate-buffered?
The pH of the Krebs-Ringer Solution, bicarbonate-buffered is 7.3.
Is the Krebs-Ringer Solution, bicarbonate-buffered sterile?
The Krebs-Ringer Solution, bicarbonate-buffered is prepared in 18.2 megohms water and filtered through 0.22 micron filter to remove contaminants. No sterility testing is performed so it cannot be certified as sterile.
What are the concentrations of the components of Krebs-Ringer Solution, bicarbonate-buffered?
The Krebs-Ringer Solution, bicarbonate-buffered contains 120 mM sodium chloride, 5 mM potassium chloride, 2 mM calcium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5 mM D-glucose.