TA Cloning&trade; Kit, Dual Promoter, with pCR&trade;II Vector and One Shot&trade; TOP10F' Chemically Competent <i>E. coli</i>
Invitrogen™

TA Cloning™ Kit, Dual Promoter, with pCR™II Vector and One Shot™ TOP10F' Chemically Competent E. coli

The TA Cloning™ Kit Dual Promoter with pCR™II vector provides a quick, one-step cloning strategy for directly inserting a Taq-amplifiedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K20600120 Reactions
K20604040 Reactions
Catalog number K206001
Price (USD)
780.00
Each
No. of Reactions:
20 Reactions
Price (USD)
780.00
Each
The TA Cloning™ Kit Dual Promoter with pCR™II vector provides a quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into a plasmid vector. The T7 and Sp6 promoters of the pCR™II vector allow in vitro transcription of the insert to produce sense or anti-sense products. The TA Cloning Kit Dual Promoter uses the pCR™II cloning vector and ExpressLink™ T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

Features of the TA Cloning™ Kit Dual Promoter:
Fast & convenient—15-minute, room-temperature ligation
Efficient—blue/white screening and >80% clones with correct insert
Flexible—choice of kanamycin or ampicillin resistance for flexible antibiotic selection
Hassle-free—eliminates any enzymatic modifications of the PCR product
Streamlined—does not require the use of PCR primers that contain restriction sites

The pCR™II vector provides:
• 3'-T overhangs for direct ligation of Taq-amplified PCR products
• T7 and Sp6 promoters for in vitro RNA transcription and sequencing
• Versatile polylinker with flanking EcoR I sites for easy excision of inserts
• M13 forward and reverse primer sites for sequencing

How TA Cloning™ Works
Taq polymerase has a non-template-dependent activity that adds a single deoxyadenosine (A) to the 3' ends of PCR products. The linearized vector supplied in this kit has single 3' deoxythymidine (T) residues. This allows PCR inserts to ligate efficiently with the vector.

Kit Configurations
The TA Cloning™ Kit is offered in a variety of configurations: with One Shot™ INVF' Chemically Competent E. coli (K2050-01 and K2050-40), One Shot™ TOP10F' Chemically Competent E. coli (K2060-01 and K2060-40), and without competent cells (K2750-20 and K2750-40) in 20- and 40-reaction kit sizes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial or Yeast StrainTOP10F ́
Cell TypeChemically Competent E. coli
Cloning MethodTA Cloning
For Use With (Application)PCR Cloning
IncludesLinearized pCRII vector, ExpressLink T4 DNA ligase, 5X ExpressLink T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, controls, One Shot TOP10F' chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
No. of Reactions20 Reactions
Product LineOne Shot
Product TypeCloning Kit
PromoterT7, SP6
Quantity20 reactions
VectorpCRII
FormatKit
Unit SizeEach
Contents & Storage
This kit contains linearized pCR™II vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

Can I use TOP10F' competent cells for transformation of my TOPO vector that contains the ccdB gene?

Strains that contain an F plasmid, such as TOP10F', are not recommended for transformation and selection of recombinant clones in any TOPO vector containing the ccdB gene. While the F plasmid does encode the CcdA protein, which acts as an inhibitor of the CcdB gyrase-toxin protein, the half-life of the CcdA protein is shorter than that of the CcdB protein. Overexpression of the CcdB protein causes cell death when its action is not prevented by sufficient CcdA.

Have you compared in vitro transcription levels between SP6 and T7 promoters in your pCRII vectors?

No, we have not done in-house comparisons of transcription levels. It is widely known though that T7 polymerase produces more RNA than SP6 (on the order of 10-fold higher).

Citations & References (141)

Citations & References
Abstract
LMP-1, a LIM-domain protein, mediates BMP-6 effects on bone formation.
Authors:Boden SD, Liu Y, Hair GA, Helms JA, Hu D, Racine M, Nanes MS, Titus L
Journal:Endocrinology
PubMed ID:9832452
Glucocorticoids can promote osteoblast differentiation from fetal calvarial cells and bone marrow stromal cells. We recently reported that glucocorticoid specifically induced bone morphogenetic protein-6 (BMP-6), a glycoprotein signaling molecule that is a multifunctional regulator of vertebrate development. In the present study, we used fetal rat secondary calvarial cultures to determine ... More
Glycoprotein IIb Leu214Pro mutation produces glanzmann thrombasthenia with both quantitative and qualitative abnormalities in GPIIb/IIIa.
Authors:Grimaldi CM, Chen F, Wu C, Weiss HJ, Coller BS, French DL
Journal:Blood
PubMed ID:9473221
Glanzmann thrombasthenia is an inherited bleeding disorder due to a functional reduction or absence of platelet GPIIb/IIIa (alphaIIbbeta3) integrin receptors. Based on a prolonged bleeding time and absence of platelet aggregation in response to physiologic agonists, a 55-year-old white man was diagnosed as having Glanzmann thrombasthenia. The patient's platelet fibrinogen ... More
Type IV collagen is detectable in most, but not all, basement membranes of Caenorhabditis elegans and assembles on tissues that do not express it.
Authors:Graham PL, Johnson JJ, Wang S, Sibley MH, Gupta MC, Kramer JM
Journal:J Cell Biol
PubMed ID:9166416
Type IV collagen in Caenorhabditis elegans is produced by two essential genes, emb-9 and let-2, which encode alpha1- and alpha2-like chains, respectively. The distribution of EMB-9 and LET-2 chains has been characterized using chain-specific antisera. The chains colocalize, suggesting that they may function in a single heterotrimeric collagen molecule. Type ... More
Distribution and cloning of eukaryotic mRNAs by means of differential display: refinements and optimization.
Authors:Liang P, Averboukh L, Pardee AB
Journal:Nucleic Acids Res
PubMed ID:8341601
Differential display has been developed as a tool to detect and characterize altered gene expression in eukaryotic cells. The basic principle is to systematically amplify messenger RNAs and then distribute their 3' termini on a denaturing polyacrylamide gel. Here we provide methodological details and examine in depth the specificity, sensitivity ... More
Cloning and characterization of a naturally occurring antisense RNA to human thymidylate synthase mRNA.
Authors:Dolnick BJ
Journal:Nucleic Acids Res
PubMed ID:8493092
Based upon reverse transcription and polymerase chain reaction results with human KB cell RNA, a cDNA (i.e., 3'rTS1, 1557 nt) with complementarity to thymidylate synthase mRNA was cloned and sequenced. Northern blot analysis showed that 3'rTS1 corresponded to a cytoplasmic 1.8 kb RNA found in several tumor cell lines. The ... More