Thermo Scientific Chemicals

N(epsilon)-Benzyloxycarbonyl-L-lysine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: L09744.06
5 g, Each
Thermo Scientific Chemicals

N(epsilon)-Benzyloxycarbonyl-L-lysine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: L09744.06
5 g, Each
Quantity
Catalog number: L09744.06
also known as L09744-06
Price (USD)
Price: 106.00
Online price: 90.65
Your price:
Quantity
-

Chemical Identifiers

CAS
27894-50-4
IUPAC Name
methyl (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoate hydrochloride
Molecular Formula
C15H23ClN2O4
InChI Key
QPNJISLOYQGQTI-ZOWNYOTGSA-N
SMILES
Cl.COC(=O)[C@@H](N)CCCCNC(=O)OCC1=CC=CC=C1
Appearance (Color)
White to pale yellow
Assay (Titration ex Chloride)
≥94.0 to ≤106.0%
Optical Rotation
+15.0° ± 1.0° (c=2 in Water)
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text