Thermo Scientific Chemicals

N-Benzylglycine ethyl ester, 97%, Thermo Scientific Chemicals

Catalog number: L15496.14
25 g, Each
Thermo Scientific Chemicals

N-Benzylglycine ethyl ester, 97%, Thermo Scientific Chemicals

Catalog number: L15496.14
25 g, Each
Quantity
Catalog number: L15496.14
also known as L15496-14
Price (USD)
Price: 66.40
Online price: 57.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6436-90-4
IUPAC Name
ethyl 2-(benzylamino)acetate
Molecular Formula
C11H15NO2
InChI Key
ULOLIZHBYWAICY-UHFFFAOYSA-N
SMILES
CCOC(=O)CNCC1=CC=CC=C1
Form
Liquid
Refractive Index
1.5030-1.5090 @ 20°C (non-U.S. specification)
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text