Search Thermo Fisher Scientific
Reconstitute with 1 ml of distilled water. After reconstitution, store product undiluted at 4°C in the dark.
For FACS analysis, use 10ul of the suggested working dilution to label 1x10^6 cells in 100ul.
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter's Syndrome.
Protein Aliases: CD antigen CD5; CD5; CD5 antigen p56-62; fCD5; Pan T cell