Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 19
Keep as concentrated solution.
Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (92%), Japanese Medaka (88%), Xenopus laevis (90%), Cat (100%), Pig (98%), Chicken (94%), Sheep (100%), Rhesus Monkey (100%), Bovine (100%).
Positive Control: NT2D1, PC-3, U87-MG, SK-N-SH, human ESC, zebrafish eye.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delta EF2a; MCOPS3 (Microphthalmia Syndromic type 3); sex determining region Y-box 2; SOX 2; SRY (sex determining region Y) box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY related HMG box 2; SRY-box containing gene 2; SRY-related HMG-box gene 2; Transcription factor SOX-2; transcription factor SOX2
Gene Aliases: ANOP3; cb236; lcc; MCOPS3; RGD1565646; Sox-2; SOX2; soxp; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389
UniProt ID: (Human) P48431, (Mouse) P48432, (Zebrafish) Q6P0E1
Entrez Gene ID: (Human) 6657, (Rat) 499593, (Mouse) 20674, (Zebrafish) 378723
Molecular Function: HMG box transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support