Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Connections | USB |
Wash/Waste Bottle Capacity | 2 L |
Description | 2L wash bottle B, Wellwash Versa |
Bottles | Wash Bottle (2 L) |
Plate Type | 96-well Plate, 384-well Plate |
Type | Accessory |
For Use With (Equipment) | Wellwash Versa Microplate Washer |
Includes | Cap with Tubing and Liquid Level Sensors |
Automation Interface | Yes (USB) |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
N10808 | Each | 2L wash bottle B, Wellwash Versa | 415.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Easy and convenient to use
Reliable and secure performance
Optimal washing performance
Added versatility
Includes:
2x2L wash bottles, 1x2L rinse bottle, 1x4L waste bottle, two 1x8 wash heads, aerosol cover
Recommended for:
ELISA-based assays, cell washing