Poly-HRP Dilution Buffer
Thermo Scientific™

Poly-HRP Dilution Buffer

Thermo Scientific Pierce Poly-HRP Dilution Buffer is optimized to ensure optimal performance with Poly-HRP Streptavidin.Features of Poly-HRP Dilution Buffer:• Formulation:Read more
Have Questions?
Catalog NumberQuantity
N500100 mL
Catalog number N500
Price (USD)
150.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
150.00
Each
Thermo Scientific Pierce Poly-HRP Dilution Buffer is optimized to ensure optimal performance with Poly-HRP Streptavidin.

Features of Poly-HRP Dilution Buffer:

• Formulation: 1% biotin-free casein in a PBS buffer solution.
• Application: Dilution of Poly-HRP Streptavidin (Product # N200): Gently invert bottle and swirl to mix before use. Do not vortex. Use at full strength. Refer to Poly-HRP Streptavidin Certificate of Analysis for recommended dilution ranges. Dilute Poly-HRP Streptavidin immediately before use.

Related Products
Poly-HRP Streptavidin
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialDilution Buffer
Recommended StorageShipped on cold packs. Store at 2°C to 8°C immediately upon receipt.
Physical FormLiquid
Quantity100 mL
Unit SizeEach