Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Remel CTA with 1% Sucrose (Cystine Trypticase Agar) is used for determination of fermentation reactions by fastidious microorganisms, such as Neisseria, pneumococci, streptococci and non-spore forming anaerobes.
Description | CTA w/1% Sucrose |
Format | 15mm x 103mm Tube |
Product Type | Agar |
Type | Agar |
Quantity | 20 x 4 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R060660 | Each | 20 x 4 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel CTA with 1% Sucrose (Cystine Trypticase Agar) is used for determination of fermentation reactions by fastidious microorganisms, such as Neisseria, pneumococci, streptococci and non-spore forming anaerobes.