Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Selectively cultivate and isolate pathogenic fungi with Thermo Scientific™ Remel™ SABHI™ Agar w/Chloramphenicol, Cycloheximide. In 1967, Gorman developed SABHI Agar by combining Sabouraud Dextrose Agar and Brain Heart Infusion Agar. This combined formulation increased recovery of pathogenic fungi1. Selectivity can be achieved by the addition of different antibiotics. Sabouraud Dextrose Agar helps in the cultivation of dermatophytes and Brain Heart Infusion Agar helps to isolate fungi from clinical specimens2.
Description | SABHI Agar w/Chloramphenicol, Cycloheximide (Sabouraud Dextrose w/Brain Heart Infusion Agar) |
Format | 20mm x 113mm Tube |
Product Type | Agar |
Quantity | 100/Pk. |
Unit Size | Each |
Description | SABHI Agar w/Chloramphenicol, Cycloheximide (Sabouraud Dextrose w/Brain Heart Infusion Agar) |
Format | Monoplate Deep Pour |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R08697 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R01754 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
SABHI Agar w/Chloramphenicol, Cycloheximide is a solid medium recommended for use in qualitative procedures for cultivation and isolation of pathogenic fungi.
This medium is made selective by adding chloramphenicol and cycloheximide that inhibit a range of gram-positive and gram-negative bacteria and saprophytic fungi respectively.
SABHI Agar w/Chloramphenicol, Cycloheximide contains dextrose, casein peptone, meat peptone, brain heart infusion solids, sodium chloride, disodium phosphate, agar, demineralized water, chloramphenicol and cycloheximide. Dextrose serves as the carbohydrate source. Sheep blood provide essential growth factors for fastidious fungi. Chloramphenicol and gentamicin inhibit gram-positive and gram-negative bacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.