Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Identify, cultivate and enumerate yeast and molds with Thermo Scientific™ Remel™ Potato Dextrose Agar. Recommended by American Public Health Association (APHA) and Association of Official Analytical Chemists (AOAC), for plate count of yeasts and molds in the inspection of dairy products and foods1,2,3. This medium is used to prepare slide culture of fungi to stimulate the spores4,5. It is used to maintain stock cultures of dermatophytes. Rebel and Taplin used this medium to differentiate atypical dermatophytes based on the pigment produced6.
Certifications/Compliance | Industrial Reference: BAM, COMPF, USP. |
Description | Potato Dextrose Agar pH 5.6 |
Format | G - PT |
Product Type | Agar |
Quantity | 100/Pk. |
Unit Size | Each |
Certifications/Compliance | Industrial Reference: BAM, COMPF, USP. |
Description | Potato Dextrose Agar pH 5.6 |
Format | Monoplate Deep Pour |
Product Type | Agar |
Quantity | 100/Pk. |
Unit Size | Each |
Certifications/Compliance | Industrial Reference: BAM, COMPF, USP. |
Description | Potato Dextrose Agar pH 5.6 |
Format | Monoplate Deep Pour |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Certifications/Compliance | Industrial Reference: BAM, COMPF, USP. |
Description | Potato Dextrose Agar pH 5.6 |
Format | F - slant |
Product Type | Agar |
Quantity | 100/Pk. |
Unit Size | Each |
Certifications/Compliance | Industrial Reference: BAM, COMPF, USP. |
Description | Potato Dextrose Agar pH 5.6 |
Format | F - slant |
Product Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Certifications/Compliance | Industrial Reference: BAM, COMPF, USP. |
Description | Potato Dextrose Agar pH 5.6 |
Format | G - PT |
Product Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Certifications/Compliance | Industrial Reference: BAM, COMPF, USP. |
Description | Potato Dextrose Agar pH 5.6 |
Format | BB - 500mL |
Product Type | Agar |
Quantity | 10 x 500 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R09654 | Each | 100/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R01693 | Each | 100/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R01695 | Each | 10/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08650 | Each | 100/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08652 | Each | 20/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R09656 | Each | 20/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R112535 | Each | 10 x 500 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Identify, cultivate and enumerate yeast and molds using the solid medium Potato Dextrose Agar with pH 5.6.
The infusion from the potatoes provides a nutritious base for luxuriant development of most fungi. Dextrose added in the medium serves as a growth stimulant.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.