Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Remel TAT Broth (w/Polysorbate 20) is used for sterility testing of highly viscous substances, such as salves, ointments and cosmetics.
Description | TAT Broth, w/Polysorbate 20 |
Format | WMB Lug - 90mL |
Product Type | Microbiology Media |
Quantity | 10 x 90 mL |
Unit Size | Each |
Description | TAT Broth, w/Polysorbate 20 |
Format | PB -1000mL |
Product Type | Microbiology Media |
Quantity | 1000 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R112611 | Each | 10 x 90 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R112612 | Each | 1000 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel TAT Broth (w/Polysorbate 20) is used for sterility testing of highly viscous substances, such as salves, ointments and cosmetics.