Product Image
Thermo Scientific™
Remel™ TAT Broth
Thermo Scientific™ Remel TAT Broth (w/Polysorbate 20) is used for sterility testing of highly viscous substances, such as salves, ointments and cosmetics.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R1126121000 mL
R11261110 x 90 mL
Catalog number R112612
Price (USD)
-
Quantity:
1000 mL
Thermo Scientific™ Remel TAT Broth (w/Polysorbate 20) is used for sterility testing of highly viscous substances, such as salves, ointments and cosmetics.
Not intended for IVD use.
Specifications
DescriptionTAT Broth, w/Polysorbate 20
FormatPB -1000mL
Quantity1000 mL
Product TypeBroth
Unit SizeEach