Product References
IkappaBalpha is required for marginal zone B cell lineage development.
European journal of immunology
Ellinghaus U,Rupec RA,Pabst O,Ignatius R,Förster R,Dörken B,Jundt F
RM2604 was used in flow cytometry to investigate the role of nuclear factor-kappaB signaling in the development of marginal zone B cells
Fri Aug 01 00:00:00 EDT 2008
The earliest thymic progenitors in adults are restricted to T, NK, and dendritic cell lineage and have a potential to form more diverse TCRbeta chains than fetal progenitors.
Journal of immunology (Baltimore, Md. : 1950)
Lu M,Tayu R,Ikawa T,Masuda K,Matsumoto I,Mugishima H,Kawamoto H,Katsura Y
RM2604 was used in flow cytometry to study T cell progenitors in the adult thymus
Tue Nov 01 00:00:00 EST 2005
Effect of thermal injury on splenic myelopoiesis.
Shock (Augusta, Ga.)
Noel JG,Guo X,Wells-Byrum D,Schwemberger S,Caldwell CC,Ogle CK
RM2604 was used in flow cytometry to measure the number of splenic myeloid progenitors and their progeny after burn injury
Tue Feb 01 00:00:00 EST 2005
Immunoglobulin and cytokine production from spleen lymphocytes is modulated in C57BL/6J mice by dietary cis-9, trans-11 and trans-10, cis-12 conjugated linoleic acid.
The Journal of nutrition
Yamasaki M,Chujo H,Hirao A,Koyanagi N,Okamoto T,Tojo N,Oishi A,Iwata T,Yamauchi-Sato Y,Yamamoto T,Tsutsumi K,Tachibana H,Yamada K
RM2604 was used in flow cytometry to study the effect of dietary cis-9, trans- and trans-0, cis-2 conjugated linoleic acid on immunoglobulin and cytokine production from spleen lymphocytes in C57BL/6J mice.
Sat Mar 01 00:00:00 EST 2003
Growth hormone accelerates immune recovery following allogeneic T-cell-depleted bone marrow transplantation in mice.
Experimental hematology
Chen BJ,Cui X,Sempowski GD,Chao NJ
RM2604 was used in flow cytometry to test if recombinant human growth hormone promotes immune recovery after allogeneic T-cell-depleted bone marrow transplantation.
Wed Oct 01 00:00:00 EDT 2003
Suppression of lymphoma and epithelial malignancies effected by interferon gamma.
The Journal of experimental medicine
Street SE,Trapani JA,MacGregor D,Smyth MJ
RM2604 was used in flow cytometry to study knock out mice to identify genes that suppress lymphomagenesis.
Mon Jul 01 00:00:00 EDT 2002
Long-term survival of corneal allografts is dependent on intact CD1d-reactive NKT cells.
Journal of immunology (Baltimore, Md. : 1950)
Sonoda KH,Taniguchi M,Stein-Streilein J
RM2604 was used in flow cytometry to test if the survival of a cornea graft is dependent on CD1d-reactive NKT cells.
Fri Feb 15 00:00:00 EST 2002
Naive B lymphocytes undergo homeostatic proliferation in response to B cell deficit.
Journal of immunology (Baltimore, Md. : 1950)
Cabatingan MS,Schmidt MR,Sen R,Woodland RT
RM2604 was used in flow cytometry to investigate homeostatic B cell proliferation.
Sun Dec 15 00:00:00 EST 2002
Cell-intrinsic effects of non-MHC NOD genes on dendritic cell generation in vivo.
International immunology
Prasad SJ,Goodnow CC
RM2604 was used in flow cytometry to characterize the differences in dendritic cell generation in vivo caused by non-MHC NOD genes.
Sat Jun 01 00:00:00 EDT 2002
SH2-B is required for both male and female reproduction.
Molecular and cellular biology
Ohtsuka S,Takaki S,Iseki M,Miyoshi K,Nakagata N,Kataoka Y,Yoshida N,Takatsu K,Yoshimura A
RM2604 was used in Flow cytometry/Cell sorting to study the role of SH2-B in the insulin-like growth factor-mediated reproductive pathway in mice.
Wed May 01 00:00:00 EDT 2002
Immature multipotent hemopoietic progenitors lacking long-term bone marrow-reconstituting activity in the aorta-gonad-mesonephros region of murine day 10 fetuses.
Journal of immunology (Baltimore, Md. : 1950)
Ohmura K,Kawamoto H,Lu M,Ikawa T,Ozaki S,Nakao K,Katsura Y
RM2604 was used in flow cytometry to examine multipotent progenitors from early fetuses.
Thu Mar 01 00:00:00 EST 2001
Inhibition of thymidine synthesis by folate analogues induces a Fas-Fas ligand-independent deletion of superantigen-reactive peripheral T cells.
International immunology
Izeradjene K,Revillard JP,Genestier L
RM2604 was used in flow cytometry to assess the effects of folate analogues on superantigen-reactive peripheral T cells in vivo.
Mon Jan 01 00:00:00 EST 2001
Perforin-mediated cytotoxicity is critical for surveillance of spontaneous lymphoma.
The Journal of experimental medicine
Smyth MJ,Thia KY,Street SE,MacGregor D,Godfrey DI,Trapani JA
RM2604 was used in flow cytometry to test if pfp-deficient mice are more susceptible to cancer.
Mon Sep 04 00:00:00 EDT 2000
Characterization of peripheral regulatory CD4+ T cells that prevent diabetes onset in nonobese diabetic mice.
Journal of immunology (Baltimore, Md. : 1950)
Lepault F,Gagnerault MC
RM2604 was used in flow cytometry to determine the phenotype and function of T cells that prevent diabetes.
Sat Jan 01 00:00:00 EST 2000
CD1-reactive natural killer T cells are required for development of systemic tolerance through an immune-privileged site.
The Journal of experimental medicine
Sonoda KH,Exley M,Snapper S,Balk SP,Stein-Streilein J
RM2604 was used in flow cytometry to investigate the role of NKT cells in the development of tolerance.
Mon Nov 01 00:00:00 EST 1999
Emergence of T cell progenitors without B cell or myeloid differentiation potential at the earliest stage of hematopoiesis in the murine fetal liver.
Journal of immunology (Baltimore, Md. : 1950)
Kawamoto H,Ohmura K,Fujimoto S,Katsura Y
RM2604 was used in flow cytometry to assess if progenitors colonizing the thymus are T cell lineage restricted.
Mon Mar 01 00:00:00 EST 1999
Gene therapy of experimental autoimmune thyroiditis by in vivo administration of plasmid DNA coding for Fas ligand.
Journal of immunology (Baltimore, Md. : 1950)
Batteux F,Tourneur L,Trebeden H,Charreire J,Chiocchia G
RM2604 was used in flow cytometry to study FasL expression on thyrocytes.
Fri Jan 01 00:00:00 EST 1999
Emergence of T, B, and myeloid lineage-committed as well as multipotent hemopoietic progenitors in the aorta-gonad-mesonephros region of day 10 fetuses of the mouse.
Journal of immunology (Baltimore, Md. : 1950)
Ohmura K,Kawamoto H,Fujimoto S,Ozaki S,Nakao K,Katsura Y
RM2604 was used in flow cytometry to investigate the aorta-gonad-mesonephros region as a source of hemopoietic progenitors.
Mon Nov 01 00:00:00 EST 1999
Repression of B7.2 on self-reactive B cells is essential to prevent proliferation and allow Fas-mediated deletion by CD4(+) T cells.
The Journal of experimental medicine
Rathmell JC,Fournier S,Weintraub BC,Allison JP,Goodnow CC
RM2604 was used in flow cytometry to report that B7.2 repression is critical to eliminate autoreactive B cells by Fas.
Mon Aug 17 00:00:00 EDT 1998
The IL-1 receptor-related T1 antigen is expressed on immature and mature mast cells and on fetal blood mast cell progenitors.
Journal of immunology (Baltimore, Md. : 1950)
Moritz DR,Rodewald HR,Gheyselinck J,Klemenz R
RM2604 was used in flow cytometry to assess the expression of the T1 surface antigen in murine hemopoietic organs.
Sun Nov 01 00:00:00 EST 1998
Immunologic memory induced by a glycoconjugate vaccine in a murine adoptive lymphocyte transfer model.
Infection and immunity
Guttormsen HK,Wetzler LM,Finberg RW,Kasper DL
RM2604 was used in flow cytometry to develop a mouse model to study the ability of a glycoprotein conjugate vaccine to induce immunologic memory for the polysaccharide moiety.
Fri May 01 00:00:00 EDT 1998
Outer periarteriolar lymphoid sheath arrest and subsequent differentiation of both naive and tolerant immunoglobulin transgenic B cells is determined by B cell receptor occupancy.
The Journal of experimental medicine
Cook MC,Basten A,Fazekas de St Groth B
RM2604 was used in flow cytometry to determine the localization of antigen-pulsed B cells in the spleen.
Fri Aug 29 00:00:00 EDT 1997
Self-reactive B cells are not eliminated or inactivated by autoantigen expressed on thyroid epithelial cells.
The Journal of experimental medicine
Akkaraju S,Canaan K,Goodnow CC
RM2604 was used in flow cytometry to investigate how humoral tolerance is maintained in the presence of organ-specific membrane antigens.
Mon Dec 15 00:00:00 EST 1997
Surface antigen expression in spleen cells of C57B1/6 mice during ageing: influence of sex and parity.
Clinical and experimental immunology
Barrat F,Lesourd BM,Louise A,Boulouis HJ,Vincent-Naulleau S,Thibault D,Sanaa M,Neway T,Pilet CH
RM2604 was used in flow cytometry to test if pregnancies and gender influence the cellar changes observed during ageing in mice.
Sat Mar 01 00:00:00 EST 1997
Exclusive development of T cell neoplasms in mice transplanted with bone marrow expressing activated Notch alleles.
The Journal of experimental medicine
Pear WS,Aster JC,Scott ML,Hasserjian RP,Soffer B,Sklar J,Baltimore D
RM2604 was used in flow cytometry to report that TAN1 is an oncoprotein.
Wed May 01 00:00:00 EDT 1996
Isolation of the most immature population of murine fetal thymocytes that includes progenitors capable of generating T, B, and myeloid cells.
The Journal of experimental medicine
Hattori N,Kawamoto H,Katsura Y
RM2604 was used in flow cytometry to report that FcR is a useful marker to isolate the most immature population of murine fetal thymic cells.
Fri Nov 01 00:00:00 EST 1996
Decreased CD4-CD8- TCR-alpha beta + cells in lpr/lpr mice lacking beta 2-microglobulin.
Journal of immunology (Baltimore, Md. : 1950)
Mixter PF,Russell JQ,Durie FH,Budd RC
RM2604 was used in flow cytometry to test the requirement for CD8 to interact with class I MHC molecules to promote CD4-CD8- T cells in lpr/lpr mice.
Wed Mar 01 00:00:00 EST 1995
Development and function of T cells in mice with a disrupted CD2 gene.
The EMBO journal
Killeen N,Stuart SG,Littman DR
RM2604 was used in Flow cytometry/Cell sorting to study the in vivo function of CD2.
Tue Dec 01 00:00:00 EST 1992