Thermo Scientific™

Smart-Vue™ Receivers

Catalog number: SV103-500-LSB
Have Questions?
Thermo Scientific™

Smart-Vue™ Receivers

Catalog number: SV103-500-LSB
Have Questions?
Choose from an assortment of Thermo Scientific™ Smart-Vue™ receivers including receiver packages that include power supply and software for your wireless monitoring solution.
 
Catalog Number
SV103-500-LSB
Unit Size
Each
Description
USB 1.0 or 2.0 25mW Receiver w/client server software
Operating Frequency
915 MHz
Includes
Velcro, CD-ROM with Smart-Vue client software, software drivers and user manuals
Price (USD)
Full specifications
TypeReceiver, 915MHz
DescriptionUSB 1.0 or 2.0 25mW Receiver w/client server software
IncludesVelcro, CD-ROM with Smart-Vue client software, software drivers and user manuals
For Use With (Application)Cold Storage
Operating Frequency915 MHz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RS232 25/500mW Receiver w/universal power supply
Operating Frequency
915 MHz
Includes
Universal power supply
Price (USD)
Full specifications
TypeRepeater Module, 915MHz
DescriptionRS232 25/500mW Receiver w/universal power supply
IncludesUniversal power supply
For Use With (Application)Cold Storage
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RS232 25/500mW Receiver w/universal power supply and client server software
Operating Frequency
915 MHz
Includes
Universal power supply and client server software
Price (USD)
Full specifications
TypeRepeater Module, 915MHz
DescriptionRS232 25/500mW Receiver w/universal power supply and client server software
IncludesUniversal power supply and client server software
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RS232 25mW Receiver w/universal power supply
Operating Frequency
915 MHz
Includes
Universal power supply
Price (USD)
Full specifications
TypeRepeater Module, 915MHz
DescriptionRS232 25mW Receiver w/universal power supply
IncludesUniversal power supply
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Cold Storage
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
USB 1.0 or 2.0 25mW Receiver
Operating Frequency
915 MHz
Includes
Velcro, CD-ROM with software drivers and user manuals
Price (USD)
Full specifications
TypeReceiver, 915MHz
DescriptionUSB 1.0 or 2.0 25mW Receiver
IncludesVelcro, CD-ROM with software drivers and user manuals
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
USB 1.0 or 2.0 Receiver, 25mW+SW 20 sensors
Operating Frequency
915 MHz
Includes
Velcro, CD ROM with Smart-Vue client server software 2.0 license up to 20 sensors, software drivers and user manuals.
Price (USD)
Full specifications
TypeReceiver, 915MHz
DescriptionUSB 1.0 or 2.0 Receiver, 25mW+SW 20 sensors
IncludesVelcro, CD ROM with Smart-Vue client server software 2.0 license up to 20 sensors, software drivers and user manuals.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Wireless Monitoring Solution
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
USB 1.0 or 2.0 Receiver, 25mW+SW 49 sensors
Operating Frequency
Includes
Velcro, CD ROM with Smart-Vue client server software 2.0 license up to 49 sensors, software drivers and user manuals.
Price (USD)
Full specifications
DescriptionUSB 1.0 or 2.0 Receiver, 25mW+SW 49 sensors
IncludesVelcro, CD ROM with Smart-Vue client server software 2.0 license up to 49 sensors, software drivers and user manuals.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Wireless Monitoring Solution
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Unit SizeEach
Catalog Number
Unit Size
Each
Description
USB 1.0 or 2.0 Receiver, 25mW+SW 99 sensors
Operating Frequency
915 MHz
Includes
Price (USD)
Full specifications
TypeReceiver, 915MHz
DescriptionUSB 1.0 or 2.0 Receiver, 25mW+SW 99 sensors
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Wireless Monitoring Solution
Operating Frequency915 MHz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Operating Frequency
915 MHz
Includes
Velcro, CD ROM with Smart-Vue client server software 2.0 license, unlimited sensors, software drivers and user manuals.
Price (USD)
Full specifications
IncludesVelcro, CD ROM with Smart-Vue client server software 2.0 license, unlimited sensors, software drivers and user manuals.
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Operating Frequency
915 MHz
Includes
Universal (100/240V) power supply, LAN CAT 5 cable, wall mounting holder with Velcro, CD-ROM with software drivers and user manuals
Price (USD)
Full specifications
TypeReceiver, 868MHz
IncludesUniversal (100/240V) power supply, LAN CAT 5 cable, wall mounting holder with Velcro, CD-ROM with software drivers and user manuals
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Cold Storage
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
Showing 10 of 15Show more
Catalog NumberSpecificationsUnit SizeDescriptionOperating FrequencyIncludesPrice (USD)
SV103-500-LSBFull specifications
EachUSB 1.0 or 2.0 25mW Receiver w/client server software915 MHzVelcro, CD-ROM with Smart-Vue client software, software drivers and user manualsRequest A Quote
TypeReceiver, 915MHz
DescriptionUSB 1.0 or 2.0 25mW Receiver w/client server software
IncludesVelcro, CD-ROM with Smart-Vue client software, software drivers and user manuals
For Use With (Application)Cold Storage
Operating Frequency915 MHz
Unit SizeEach
SV102-501-LSBFull specifications
EachRS232 25/500mW Receiver w/universal power supply915 MHzUniversal power supplyRequest A Quote
TypeRepeater Module, 915MHz
DescriptionRS232 25/500mW Receiver w/universal power supply
IncludesUniversal power supply
For Use With (Application)Cold Storage
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
SV102-500-LSBFull specifications
EachRS232 25/500mW Receiver w/universal power supply and client server software915 MHzUniversal power supply and client server softwareRequest A Quote
TypeRepeater Module, 915MHz
DescriptionRS232 25/500mW Receiver w/universal power supply and client server software
IncludesUniversal power supply and client server software
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
SV102-502-LSBFull specifications
EachRS232 25mW Receiver w/universal power supply915 MHzUniversal power supplyRequest A Quote
TypeRepeater Module, 915MHz
DescriptionRS232 25mW Receiver w/universal power supply
IncludesUniversal power supply
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Cold Storage
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
SV103-501-LSBFull specifications
EachUSB 1.0 or 2.0 25mW Receiver915 MHzVelcro, CD-ROM with software drivers and user manualsRequest A Quote
TypeReceiver, 915MHz
DescriptionUSB 1.0 or 2.0 25mW Receiver
IncludesVelcro, CD-ROM with software drivers and user manuals
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
SV103-507-LSBFull specifications
EachUSB 1.0 or 2.0 Receiver, 25mW+SW 20 sensors915 MHzVelcro, CD ROM with Smart-Vue client server software 2.0 license up to 20 sensors, software drivers and user manuals.Request A Quote
TypeReceiver, 915MHz
DescriptionUSB 1.0 or 2.0 Receiver, 25mW+SW 20 sensors
IncludesVelcro, CD ROM with Smart-Vue client server software 2.0 license up to 20 sensors, software drivers and user manuals.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Wireless Monitoring Solution
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
SV103-508-LSBFull specifications
EachUSB 1.0 or 2.0 Receiver, 25mW+SW 49 sensorsVelcro, CD ROM with Smart-Vue client server software 2.0 license up to 49 sensors, software drivers and user manuals.Request A Quote
DescriptionUSB 1.0 or 2.0 Receiver, 25mW+SW 49 sensors
IncludesVelcro, CD ROM with Smart-Vue client server software 2.0 license up to 49 sensors, software drivers and user manuals.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Wireless Monitoring Solution
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Unit SizeEach
SV103-509-LSBFull specifications
EachUSB 1.0 or 2.0 Receiver, 25mW+SW 99 sensors915 MHzRequest A Quote
TypeReceiver, 915MHz
DescriptionUSB 1.0 or 2.0 Receiver, 25mW+SW 99 sensors
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Wireless Monitoring Solution
Operating Frequency915 MHz
Unit SizeEach
SV103-510-LSBFull specifications
Each915 MHzVelcro, CD ROM with Smart-Vue client server software 2.0 license, unlimited sensors, software drivers and user manuals.Request A Quote
IncludesVelcro, CD ROM with Smart-Vue client server software 2.0 license, unlimited sensors, software drivers and user manuals.
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
SV104-500-LSBFull specifications
Each915 MHzUniversal (100/240V) power supply, LAN CAT 5 cable, wall mounting holder with Velcro, CD-ROM with software drivers and user manualsRequest A Quote
TypeReceiver, 868MHz
IncludesUniversal (100/240V) power supply, LAN CAT 5 cable, wall mounting holder with Velcro, CD-ROM with software drivers and user manuals
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Cold Storage
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Solutions
Operating Frequency915 MHz
Unit SizeEach
Showing 10 of 15Show more

Compatible with:

Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens

Ordering Information:

Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text