Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RLLLVTSLV VVLLWEAGAV PAPKVPIKMQ VKHWPSEQDP ENRAWGARVV EPPEKDDQLV VLFPVQKPKL LTTEEKPRGQ GRGPILPGTK AWMETEDTLG RVLSPEPDHD SLYHPPPEED QGEERPRLWV MPNHQVLLGP EEDQDHIYHP Q (2-151 aa encoded by BC029447)
Proline rich acidic protein 1 (PRAP1) is also named as PRO1195 or UPA. It may play an important role in maintaining normal growth homeostasis in epithelial cells. Mitotic arrest deficient 1 (MAD1) is a key component in mitotic checkpoint signaling. Recent study identified PRAP1 as a novel MAD1 binding partner using yeast-two hybrid screening. PRAP1 has four isoforms with MW 16-20 kDa and 10 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epididymis tissue protein Li 178; Pregnancy-specific uterine protein; Proline-rich acidic protein 1; Uterine-specific proline-rich acidic protein
Gene Aliases: PRAP1; PRO1195; UNQ608/PRO1195; UPA
UniProt ID: (Human) Q96NZ9, (Mouse) Q60874
Entrez Gene ID: (Human) 118471, (Mouse) 22264
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support