Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Description: The 1C1 monoclonal antibody reacts with the human CD131 molecule, also known as the common beta subunit (betaC). The common beta subunit associates with the specific alpha subunits of IL-3 receptor, IL-5 receptor and GM-CSF receptor to form high affinity receptors for these cytokines. These cytokine receptors are expressed by neutrophils, eosinophils, monocytes, endothelial cells, fibroblasts and hematopoietic progenitor cells and play a crucial role in growth/activation of eosinophils and in the inflammatory response.
Applications Reported: This 1C1 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This 1C1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Filtration: 0.2 µm post-manufacturing filtered.
The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta common cytokine receptor; CD antigen CD131; CD131; CDw131; colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage); colony-stimulating factor-2 receptor, beta, low-affinity; Cytokine receptor common subunit beta; GM-CSF/IL-3/IL-5 receptor common beta subunit; GM-CSF/IL-3/IL-5 receptor common beta-chain; IL-3 RB; IL-3 Receptor beta; IL3R beta; interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3 receptor, beta (high affinity)
Gene Aliases: CD131; CDw131; CSF2RB; IL3RB; IL5RB; SMDP5
UniProt ID: (Human) P32927
Entrez Gene ID: (Human) 1439
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support