Product References
Chaperone-mediated autophagy plays an important role in regulating retinal progenitor cell homeostasis.
Stem cell research & therapy
Jin C,Ou Q,Chen J,Wang T,Zhang J,Wang Z,Wang Y,Tian H,Xu JY,Gao F,Wang J,Li J,Lu L,Xu GT
Fri Apr 01 00:00:00 EDT 2022
RHEB/mTOR hyperactivity causes cortical malformations and epileptic seizures through increased axonal connectivity.
PLoS biology
Proietti Onori M,Koene LMC,Schäfer CB,Nellist M,de Brito van Velze M,Gao Z,Elgersma Y,van Woerden GM
Sat May 01 00:00:00 EDT 2021
PPT1 Reduction Contributes to Erianin-Induced Growth Inhibition in Oral Squamous Carcinoma Cells.
Frontiers in cell and developmental biology
Luo Q,Li X,Gan G,Yang M,Chen X,Chen F
Sat Jul 16 00:00:00 EDT 2022
MYH9 is crucial for stem cell-like properties in non-small cell lung cancer by activating mTOR signaling.
Cell death discovery
Chen M,Sun LX,Yu L,Liu J,Sun LC,Yang ZH,Shu X,Ran YL
Mon Oct 11 00:00:00 EDT 2021
Rheb1 promotes glucose-stimulated insulin secretion in human and mouse β-cells by upregulating GLUT expression.
Metabolism: clinical and experimental
Yang Y,Cai Z,Pan Z,Liu F,Li D,Ji Y,Zhong J,Luo H,Hu S,Song L,Yu S,Li T,Li J,Ma X,Zhang W,Zhou Z,Liu F,Zhang J
Fri Oct 01 00:00:00 EDT 2021
Synergistic antifibrotic effects of miR-451 with miR-185 partly by co-targeting EphB2 on hepatic stellate cells.
Cell death & disease
Chen X,Zhang D,Wang Y,Chen K,Zhao L,Xu Y,Jiang H,Wang S
Thu May 28 00:00:00 EDT 2020
CircRNA-100338 Is Associated With mTOR Signaling Pathway and Poor Prognosis in Hepatocellular Carcinoma.
Frontiers in oncology
Huang XY,Huang ZL,Zhang PB,Huang XY,Huang J,Wang HC,Xu B,Zhou J,Tang ZY
Thu Oct 01 00:00:00 EDT 2020
A brain somatic RHEB doublet mutation causes focal cortical dysplasia type II.
Experimental & molecular medicine
Zhao S,Li Z,Zhang M,Zhang L,Zheng H,Ning J,Wang Y,Wang F,Zhang X,Gan H,Wang Y,Zhang X,Luo H,Bu G,Xu H,Yao Y,Zhang YW
Tue Jul 23 00:00:00 EDT 2019
Ox-LDL-Induced MicroRNA-155 Promotes Autophagy in Human Endothelial Cells via Repressing the Rheb/ mTOR Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Lv J,Yang L,Guo R,Shi Y,Zhang Z,Ye J
Mon Dec 18 00:00:00 EST 2017
Ox-LDL-Induced MicroRNA-155 Promotes Autophagy in Human Endothelial Cells via Repressing the Rheb/ mTOR Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Lv J,Yang L,Guo R,Shi Y,Zhang Z,Ye J
Mon Dec 18 00:00:00 EST 2017
Rheb phosphorylation is involved in p38-regulated/activated protein kinase-mediated tumor suppression in liver cancer.
Oncology letters
Zheng M,Zang S,Xie L,Fang X,Zhang YU,Ma X,Liu J,Lin D,Huang A
Tue Sep 01 00:00:00 EDT 2015
Rheb phosphorylation is involved in p38-regulated/activated protein kinase-mediated tumor suppression in liver cancer.
Oncology letters
Zheng M,Zang S,Xie L,Fang X,Zhang YU,Ma X,Liu J,Lin D,Huang A
Tue Sep 01 00:00:00 EDT 2015
Rheb phosphorylation is involved in p38-regulated/activated protein kinase-mediated tumor suppression in liver cancer.
Oncology letters
Zheng M,Zang S,Xie L,Fang X,Zhang YU,Ma X,Liu J,Lin D,Huang A
Tue Sep 01 00:00:00 EDT 2015
Rheb phosphorylation is involved in p38-regulated/activated protein kinase-mediated tumor suppression in liver cancer.
Oncology letters
Zheng M,Zang S,Xie L,Fang X,Zhang YU,Ma X,Liu J,Lin D,Huang A
Tue Sep 01 00:00:00 EDT 2015
Crosstalk between glucocorticoid receptor and nutritional sensor mTOR in skeletal muscle.
Cell metabolism
Shimizu N,Yoshikawa N,Ito N,Maruyama T,Suzuki Y,Takeda S,Nakae J,Tagata Y,Nishitani S,Takehana K,Sano M,Fukuda K,Suematsu M,Morimoto C,Tanaka H
Wed Feb 02 00:00:00 EST 2011
Crosstalk between glucocorticoid receptor and nutritional sensor mTOR in skeletal muscle.
Cell metabolism
Shimizu N,Yoshikawa N,Ito N,Maruyama T,Suzuki Y,Takeda S,Nakae J,Tagata Y,Nishitani S,Takehana K,Sano M,Fukuda K,Suematsu M,Morimoto C,Tanaka H
Wed Feb 02 00:00:00 EST 2011