Product References
Adaptive immune responses to SARS-CoV-2 persist in the pharyngeal lymphoid tissue of children.
Nature immunology
Xu Q,Milanez-Almeida P,Martins AJ,Radtke AJ,Hoehn KB,Oguz C,Chen J,Liu C,Tang J,Grubbs G,Stein S,Ramelli S,Kabat J,Behzadpour H,Karkanitsa M,Spathies J,Kalish H,Kardava L,Kirby M,Cheung F,Preite S,Duncker PC,Kitakule MM,Romero N,Preciado D,Gitman L,Koroleva G,Smith G,Shaffer A,McBain IT,McGuire PJ,Pittaluga S,Germain RN,Apps R,Schwartz DM,Sadtler K,Moir S,Chertow DS,Kleinstein SH,Khurana S,Tsang JS,Mudd P,Schwartzberg PL,Manthiram K
47-0048-42 was used in Flow cytometry/Cell sorting to provide evidence for persistent tissue-specific immunity to SARS-CoV-2 in the upper respiratory tract of children after infection.
Sun Jan 01 00:00:00 EST 2023
Logic-gated antibody pairs that selectively act on cells co-expressing two antigens.
Nature biotechnology
Oostindie SC,Rinaldi DA,Zom GG,Wester MJ,Paulet D,Al-Tamimi K,van der Meijden E,Scheick JR,Wilpshaar T,de Jong B,Hoff-van den Broek M,Grattan RM,Oosterhoff JJ,Vignau J,Verploegen S,Boross P,Beurskens FJ,Lidke DS,Schuurman J,de Jong RN
47-0048-42 was used in Flow cytometry/Cell sorting to show that HexElect® allows selective antibody activity on target cells expressing unique, potentially unexplored combinations of surface antigens.
Sat Oct 01 00:00:00 EDT 2022
Estrogen Protects against Renal Ischemia-Reperfusion Injury by Regulating Th17/Treg Cell Immune Balance.
Disease markers
Zhang Y,Chang Y,Han Z,Ma K,Zeng X,Li L
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Tue Oct 18 00:00:00 EDT 2022
Plasma Extracellular Vesicle Subtypes May be Useful as Potential Biomarkers of Immune Activation in People With HIV.
Pathogens & immunity
Bazié WW,Boucher J,Vitry J,Goyer B,Routy JP,Tremblay C,Trottier S,Jenabian MA,Provost P,Alary M,Gilbert C
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Sat Apr 23 00:00:00 EDT 2022
Targeting Phosphatidylserine Enhances the Anti-tumor Response to Tumor-Directed Radiation Therapy in a Preclinical Model of Melanoma.
Cell reports
Budhu S,Giese R,Gupta A,Fitzgerald K,Zappasodi R,Schad S,Hirschhorn D,Campesato LF,De Henau O,Gigoux M,Liu C,Mazo G,Deng L,Barker CA,Wolchok JD,Merghoub T
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Small-molecule antagonist of VLA-4 (GW559090) attenuated neuro-inflammation by targeting Th17 cell trafficking across the blood-retinal barrier in experimental autoimmune uveitis.
Journal of neuroinflammation
Chen YH,Eskandarpour M,Zhang X,Galatowicz G,Greenwood J,Lightman S,Calder V
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Thu Feb 18 00:00:00 EST 2021
A Cross-Reactive Small Protein Binding Domain Provides a Model to Study Off-Tumor CAR-T Cell Toxicity.
Molecular therapy oncolytics
Hammill JA,Kwiecien JM,Dvorkin-Gheva A,Lau VWC,Baker C,Wu Y,Bezverbnaya K,Aarts C,Heslen CW,Denisova GF,Derocher H,Milne K,Nelson BH,Bramson JL
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Fri Jun 26 00:00:00 EDT 2020
The Potential of Nanobody-Targeted Photodynamic Therapy to Trigger Immune Responses.
Cancers
Beltrán Hernández I,Angelier ML,Del Buono D'Ondes T,Di Maggio A,Yu Y,Oliveira S
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Wed Apr 15 00:00:00 EDT 2020
Expression of CD20 after viral reactivation renders HIV-reservoir cells susceptible to Rituximab.
Nature communications
Serra-Peinado C,Grau-Expósito J,Luque-Ballesteros L,Astorga-Gamaza A,Navarro J,Gallego-Rodriguez J,Martin M,Curran A,Burgos J,Ribera E,Raventós B,Willekens R,Torrella A,Planas B,Badía R,Garcia F,Castellví J,Genescà M,Falcó V,Buzon MJ
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Fri Aug 16 00:00:00 EDT 2019
Accumulation of T-helper 22 cells, interleukin-22 and myeloid-derived suppressor cells promotes gastric cancer progression in elderly patients.
Oncology letters
Chen X,Wang Y,Wang J,Wen J,Jia X,Wang X,Zhang H
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2018
IL-6 receptor blockade corrects defects of XIAP-deficient regulatory T cells.
Nature communications
Hsieh WC,Hsu TS,Chang YJ,Lai MZ
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Wed Jan 31 00:00:00 EST 2018
Complement receptor CD46 co-stimulates optimal human CD8+ T cell effector function via fatty acid metabolism.
Nature communications
Arbore G,West EE,Rahman J,Le Friec G,Niyonzima N,Pirooznia M,Tunc I,Pavlidis P,Powell N,Li Y,Liu P,Servais A,Couzi L,Fremeaux-Bacchi V,Placais L,Ferraro A,Walsh PR,Kavanagh D,Afzali B,Lavender P,Lachmann HJ,Kemper C
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Wed Oct 10 00:00:00 EDT 2018
Ex vivo culture of human atherosclerotic plaques: A model to study immune cells in atherogenesis.
Atherosclerosis
Lebedeva A,Vorobyeva D,Vagida M,Ivanova O,Felker E,Fitzgerald W,Danilova N,Gontarenko V,Shpektor A,Vasilieva E,Margolis L
47-0048 was used in Flow cytometry/Cell sorting to establish a new ex vivo model of human atherosclerotic plaques that preserves the main cell types in plaques and the extracellular components in the context of native cytoarchitecture.
Fri Dec 01 00:00:00 EST 2017
Hormonal vitamin D up-regulates tissue-specific PD-L1 and PD-L2 surface glycoprotein expression in humans but not mice.
The Journal of biological chemistry
Dimitrov V,Bouttier M,Boukhaled G,Salehi-Tabar R,Avramescu RG,Memari B,Hasaj B,Lukacs GL,Krawczyk CM,White JH
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Fri Dec 15 00:00:00 EST 2017
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Thu Aug 03 00:00:00 EDT 2017
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Tue Sep 12 00:00:00 EDT 2017
PD-L1 expression and prognostic impact in glioblastoma.
Neuro-oncology
Nduom EK,Wei J,Yaghi NK,Huang N,Kong LY,Gabrusiewicz K,Ling X,Zhou S,Ivan C,Chen JQ,Burks JK,Fuller GN,Calin GA,Conrad CA,Creasy C,Ritthipichai K,Radvanyi L,Heimberger AB
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2016
Tax and Semaphorin 4D Released from Lymphocytes Infected with Human Lymphotropic Virus Type 1 and Their Effect on Neurite Growth.
AIDS research and human retroviruses
Quintremil S,Alberti C,Rivera M,Medina F,Puente J,Cartier L,Ramírez E,Tanaka Y,Valenzuela MA
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
Tumor-derived exosomes regulate expression of immune function-related genes in human T cell subsets.
Scientific reports
Muller L,Mitsuhashi M,Simms P,Gooding WE,Whiteside TL
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Thu Feb 04 00:00:00 EST 2016
The HIV-1 gp120 CD4-bound conformation is preferentially targeted by antibody-dependent cellular cytotoxicity-mediating antibodies in sera from HIV-1-infected individuals.
Journal of virology
Veillette M,Coutu M,Richard J,Batraville LA,Dagher O,Bernard N,Tremblay C,Kaufmann DE,Roger M,Finzi A
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2015
Aging and cytomegalovirus infection differentially and jointly affect distinct circulating T cell subsets in humans.
Journal of immunology (Baltimore, Md. : 1950)
Wertheimer AM,Bennett MS,Park B,Uhrlaub JL,Martinez C,Pulko V,Currier NL,Nikolich-Žugich D,Kaye J,Nikolich-Žugich J
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Sat Mar 01 00:00:00 EST 2014
CD4-CCR5 interaction in intracellular compartments contributes to receptor expression at the cell surface.
Blood
Achour L,Scott MG,Shirvani H,Thuret A,Bismuth G,Labbé-Jullié C,Marullo S
Published figure using CD4 monoclonal antibody (Product # 47-0048-42) in Flow Cytometry
Thu Feb 26 00:00:00 EST 2009