Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
May be involved in vasopressin signaling in the kidney. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat and SAM domain-containing protein 3
Gene Aliases: 2700067D09Rik; ANKS3; C81345; KIAA1977; mKIAA1977
UniProt ID: (Human) Q6ZW76, (Mouse) Q9CZK6
Entrez Gene ID: (Human) 124401, (Mouse) 72615
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support