Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This antibody will not cross-react with hamster, mouse, Xenopus, rat or chicken.
Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-8; cytokeratin 8; Cytokeratin-8; Cytokeratin-A; K8; KERA; keratin 8, type II; Keratin, type II cytoskeletal 8; Keratin-8; type II cytokeratin A (no.8); Type-II keratin Kb8
Gene Aliases: CARD2; CK-8; CK8; CYK8; K2C8; K8; KO; KRT2-8; KRT8
UniProt ID: (Bovine) P05786, (Human) P05787
Entrez Gene ID: (Rabbit) 100348891, (Bovine) 281269, (Pig) 100152077, (Human) 3856, (Sheep) 101110495
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support