Product References
Litmus-Body: A Molecularly Targeted Sensor for Cell-Surface pH Measurements.
ACS sensors
Kuo JC,Goudge MC,Metzloff AE,Huang LT,Colville MJ,Park S,Zipfel WR,Paszek MJ
MA5-12880 was used in Immunocytochemistry-immunoflourescence to show that the Litmus-body can be successfully targeted to IgG antibodies and provide localized pH measurements in the vicinity of specific cell surface components, as well as following their transit through the endocytic pathway.
Fri Jun 26 00:00:00 EDT 2020
Sensitization of EGFR Wild-Type Non-Small Cell Lung Cancer Cells to EGFR-Tyrosine Kinase Inhibitor Erlotinib.
Molecular cancer therapeutics
Raimbourg J,Joalland MP,Cabart M,de Plater L,Bouquet F,Savina A,Decaudin D,Bennouna J,Vallette FM,Lalier L
MA5-12880 was used in Flow cytometry/Cell sorting to study the effects of cisplatin treatment on the EGFR pathway in non-small cell lung cancer and the functional consequences in animal models of patient-derived xenografts.
Tue Aug 01 00:00:00 EDT 2017
IKKα regulates human keratinocyte migration through surveillance of the redox environment.
Journal of cell science
Lisse TS,Rieger S
MA512880 was used in immunocytochemistry to study H2O2-dependent wound repair mechanisms in human keratinocytes
Wed Mar 01 00:00:00 EST 2017
Interaction of EGFR to δ-catenin leads to δ-catenin phosphorylation and enhances EGFR signaling.
Scientific reports
He Y,Ryu T,Shrestha N,Yuan T,Kim H,Shrestha H,Cho YC,Seo YW,Song WK,Kim K
Published figure using EGFR monoclonal antibody (Product # MA5-12880) in Western Blot
Wed Feb 17 00:00:00 EST 2016
Interaction of EGFR to δ-catenin leads to δ-catenin phosphorylation and enhances EGFR signaling.
Scientific reports
He Y,Ryu T,Shrestha N,Yuan T,Kim H,Shrestha H,Cho YC,Seo YW,Song WK,Kim K
Published figure using EGFR monoclonal antibody (Product # MA5-12880) in Western Blot
Wed Feb 17 00:00:00 EST 2016
PHD4 stimulates tumor angiogenesis in osteosarcoma cells via TGF-α.
Molecular cancer research : MCR
Klotzsche-von Ameln A,Prade I,Grosser M,Kettelhake A,Rezaei M,Chavakis T,Flamme I,Wielockx B,Breier G
MA5-12880 was used in blocking or activating experiment to study the role of TGF-alpha in the mechanism by which prolyl hydroxylase-4 promotes osteosarcoma cell tumor angiogenesis
Fri Nov 01 00:00:00 EDT 2013
In vitro perforation of human epithelial carcinoma cell with antibody-conjugated biodegradable microspheres illuminated by a single 80 femtosecond near-infrared laser pulse.
International journal of nanomedicine
Terakawa M,Tsunoi Y,Mitsuhashi T
MA5-12880 was used in immunocytochemistry to study the ability of antibody-conjugated biodegradable microspheres excited by a single 80 femtosecond near-IR laser pulse to perforate the cell membrane of epithelial carcinoma cells
Mon Dec 10 00:00:00 EST 2012
Epidermal growth factor receptor as a biomarker for cervical cancer.
Annals of oncology : official journal of the European Society for Medical Oncology
Soonthornthum T,Arias-Pulido H,Joste N,Lomo L,Muller C,Rutledge T,Verschraegen C
MA5-12880 was used in immunohistochemistry to investigate the expression of epidermal growth factor receptor in cervical cancer
Sat Oct 01 00:00:00 EDT 2011
The β-catenin/Tcf4/survivin signaling maintains a less differentiated phenotype and high proliferative capacity of human corneal epithelial progenitor cells.
The international journal of biochemistry & cell biology
Lu R,Bian F,Zhang X,Qi H,Chuang EY,Pflugfelder SC,Li DQ
MA5-12880 was used in immunocytochemistry to study the role of the beta-catenin-Tcf4-survivin signaling pathway in human corneal epithelial progenitor cells
Sun May 01 00:00:00 EDT 2011
An EGFR autocrine loop encodes a slow-reacting but dominant mode of mechanotransduction in a polarized epithelium.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Kojic N,Chung E,Kho AT,Park JA,Huang A,So PT,Tschumperlin DJ
MA5-12880 was used in blocking or activating experiment to investigate lateral intercellular space deformation between bronchial epithelial cells in response to compressive stress
Sat May 01 00:00:00 EDT 2010
The antimicrobial peptide LL37 induces the migration of human pulp cells: a possible adjunct for regenerative endodontics.
Journal of endodontics
Kajiya M,Shiba H,Komatsuzawa H,Ouhara K,Fujita T,Takeda K,Uchida Y,Mizuno N,Kawaguchi H,Kurihara H
MA5-12880 was used in blocking or activating experiment to investigate the influence of antimicrobial peptide LL37 on human pulp cell migration
Tue Jun 01 00:00:00 EDT 2010
Nano-bio-chip sensor platform for examination of oral exfoliative cytology.
Cancer prevention research (Philadelphia, Pa.)
Weigum SE,Floriano PN,Redding SW,Yeh CK,Westbrook SD,McGuff HS,Lin A,Miller FR,Villarreal F,Rowan SD,Vigneswaran N,Williams MD,McDevitt JT
MA5-12880 was used in immunocytochemistry to develop a nano-bio-chip sensor to detect biomarkers of early oral tumorigenesis
Thu Apr 01 00:00:00 EDT 2010
Diminished survival of human cytotrophoblast cells exposed to hypoxia/reoxygenation injury and associated reduction of heparin-binding epidermal growth factor-like growth factor.
American journal of obstetrics and gynecology
Leach RE,Kilburn BA,Petkova A,Romero R,Armant DR
MA5-12880 was used in blocking or activating experiment to study the role of HB-EGF as a survival factor in human cytotrophoblast cells exposed to hypoxia/reoxygenation injury
Tue Apr 01 00:00:00 EDT 2008
Epidermal growth factor-like growth factors prevent apoptosis of alcohol-exposed human placental cytotrophoblast cells.
Biology of reproduction
Wolff GS,Chiang PJ,Smith SM,Romero R,Armant DR
MA5-12880 was used in blocking or activating experiment to examine the anti-apoptotic effect of epidermal growth factor-like growth factors in human placental cytotrophoblast cells
Sun Jul 01 00:00:00 EDT 2007
Involvement of membrane-type bile acid receptor M-BAR/TGR5 in bile acid-induced activation of epidermal growth factor receptor and mitogen-activated protein kinases in gastric carcinoma cells.
Biochemical and biophysical research communications
Yasuda H,Hirata S,Inoue K,Mashima H,Ohnishi H,Yoshiba M
MA5-12880 was used in western blot to investigate the role of membrane-type bile acid receptor M-BAR/TGR5 in bile acid-induced activation of EGFR and MAPK in gastric carcinoma cells
Fri Mar 02 00:00:00 EST 2007
Human trophoblast survival at low oxygen concentrations requires metalloproteinase-mediated shedding of heparin-binding EGF-like growth factor.
Development (Cambridge, England)
Armant DR,Kilburn BA,Petkova A,Edwin SS,Duniec-Dmuchowski ZM,Edwards HJ,Romero R,Leach RE
MA5-12880 was used in blocking or activating experiment to study the role of metalloproteinase-mediated shedding of HB-EGF-like growth factor in human trophoblast survival at low oxygen concentrations
Wed Feb 01 00:00:00 EST 2006
Receptor-selective retinoids inhibit the growth of normal and malignant breast cells by inducing G1 cell cycle blockade.
Breast cancer research and treatment
Wu K,DuPré E,Kim H,Tin-U CK,Bissonnette RP,Lamph WW,Brown PH
MA5-12880 was used in blocking or activating experiment to study the role of G1 cell cycle blockade in the inhibition of normal and cancerous breast cell gowth by synthetic, receptor selctive retionoids
Wed Mar 01 00:00:00 EST 2006
Inhibition of xenograft tumor growth and down-regulation of ErbB receptors by an antibody directed against Lewis Y antigen.
The Journal of pharmacology and experimental therapeutics
Farhan H,Schuster C,Klinger M,Weisz E,Waxenecker G,Schuster M,Sexl V,Mudde GC,Freissmuth M,Kircheis R
MA5-12880 was used in blocking or activating experiment to study the inhibition of xenograft tumor growth and down-regulation of ErbB receptors by an antibody directed against Lewis Y antigen
Fri Dec 01 00:00:00 EST 2006
Directed evolution of the epidermal growth factor receptor extracellular domain for expression in yeast.
Proteins
Kim YS,Bhandari R,Cochran JR,Kuriyan J,Wittrup KD
MA5-12880 was used in flow cytometry to express functional and properly folded EGFR extracellular domain in yeast
Wed Mar 01 00:00:00 EST 2006
The role of LIP5 and CHMP5 in multivesicular body formation and HIV-1 budding in mammalian cells.
The Journal of biological chemistry
Ward DM,Vaughn MB,Shiflett SL,White PL,Pollock AL,Hill J,Schnegelberger R,Sundquist WI,Kaplan J
MA5-12880 was used in immunocytochemistry to study the role of LIP5 and CHMP5 in multivesicular body formation and HIV-1 budding in mammalian cells
Fri Mar 18 00:00:00 EST 2005
Fine epitope mapping of anti-epidermal growth factor receptor antibodies through random mutagenesis and yeast surface display.
Journal of molecular biology
Chao G,Cochran JR,Wittrup KD
MA5-12880 was used in immunoprecipitation to map the epitopes of therapeutically important epidermal growth factor receptor antibodies
Fri Sep 10 00:00:00 EDT 2004
Autocrine extracellular signal-regulated kinase (ERK) activation in normal human keratinocytes: metalloproteinase-mediated release of amphiregulin triggers signaling from ErbB1 to ERK.
Molecular biology of the cell
Kansra S,Stoll SW,Johnson JL,Elder JT
MA5-12880 was used in western blot to investigate the mechanism of ERK autocrine activation in normal human keratinocytes
Wed Sep 01 00:00:00 EDT 2004
Domain-level antibody epitope mapping through yeast surface display of epidermal growth factor receptor fragments.
Journal of immunological methods
Cochran JR,Kim YS,Olsen MJ,Bhandari R,Wittrup KD
MA5-12880 was used in flow cytometry to study the use of yeast surface display for identifying stable folded protein domains and characterizing antibody binding epitopes
Thu Apr 01 00:00:00 EST 2004
Autoantibodies to the epidermal growth factor receptor in systemic sclerosis, lupus, and autoimmune mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Planque S,Zhou YX,Nishiyama Y,Sinha M,O'Connor-Mccourt M,Arnett FC,Paul S
MA5-12880 was used in western blot to study the auto-immune response against epidermal growth factor receptor
Sat Feb 01 00:00:00 EST 2003
Broadly distributed chemical reactivity of natural antibodies expressed in coordination with specific antigen binding activity.
The Journal of biological chemistry
Planque S,Taguchi H,Burr G,Bhatia G,Karle S,Zhou YX,Nishiyama Y,Paul S
MA5-12880 was used in EMSA to investigate the nucleophilic reactivity of natural antibodies
Fri May 30 00:00:00 EDT 2003
Heparin-binding epidermal growth factor-like growth factor: hypoxia-inducible expression in vitro and stimulation of neurogenesis in vitro and in vivo.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Jin K,Mao XO,Sun Y,Xie L,Jin L,Nishi E,Klagsbrun M,Greenberg DA
MA5-12880 was used in blocking or activating experiment to study the possible role of HB-EGF in hypoxic-ischemic induction of neurogenesis
Mon Jul 01 00:00:00 EDT 2002
Interaction between tyrosine kinase Etk and a RUN domain- and FYVE domain-containing protein RUFY1. A possible role of ETK in regulation of vesicle trafficking.
The Journal of biological chemistry
Yang J,Kim O,Wu J,Qiu Y
MA5-12880 was used in immunocytochemistry to study the interaction between tyrosine kinase Etk and RUFY1 and the potential role in vesicle trafficking
Fri Aug 16 00:00:00 EDT 2002
Differential utilization and localization of ErbB receptor tyrosine kinases in skin compared to normal and malignant keratinocytes.
Neoplasia (New York, N.Y.)
Stoll SW,Kansra S,Peshick S,Fry DW,Leopold WR,Wiesen JF,Sibilia M,Zhang T,Werb Z,Derynck R,Wagner EF,Elder JT
MA5-12880 was used in immunocytochemistry and immunoprecipitation to study the differential utilization and localization of ErbB receptor tyrosine kinases in skin compared to normal and malignant keratinocytes
Tue Dec 04 00:00:00 EST 2001