Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
MA5-15814 targets RUNX1 in indirect ELISA, IF and WB applications and shows reactivity with Human samples.
The MA5-15814 immunogen is synthesized peptide of human RUNX1. .
MA5-15814 detects RUNX1 which has a predicted molecular weight of approximately 55kDa.
RUNX1 belongs to the mammalian RUNX family of DNA binding proteins that regulate the expression of genes involved in cellular differentiation and cell cycle progression. RUNX1 is essential for hematopoietic stem cell generation in the vascular regions of the aorta, vitelline and umbilical arteries, yolk sac and placenta. RUNX1 is expressed in all tissues except brain and heart, with highest levels in thymus, bone marrow and peripheral blood.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acute myeloid leukemia 1 protein; AML-1; aml1 oncogene; AML1-EVI-1 fusion protein; CBF-alpha 2; CBF-alpha-2; Core-binding factor subunit alpha-2; core-binding factor, runt domain, alpha subunit 2; Oncogene AML-1; OTTHUMP00000108697; OTTHUMP00000108700; OTTHUMP00000108702; PEA2-alpha B; PEBP2-alpha B; Polyomavirus enhancer-binding protein 2 alpha B subunit; RUN1; Runt-related transcription factor 1; RUNX1/ZNF687 fusion; SL3-3 enhancer factor 1 alpha B subunit; SL3/AKV core-binding factor alpha B subunit; transcription factor
Gene Aliases: AML1; AML1-EVI-1; AMLCR1; CBF2alpha; CBFA2; EVI-1; PEBP2aB; PEBP2alpha; RUNX1
UniProt ID: (Human) Q01196
Entrez Gene ID: (Human) 861
Molecular Function: DNA-binding transcription factor immunoglobulin fold transcription factor Runt transcription factor gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support