Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-75193 specifically detects IL-12B p40 subunit.
PA1-75193 has been successfully used in ELISA applications.
The PA1-75193 immunogen is Recombinant IL-12B.
IL12B (IL-12 p40) encodes a subunit of interleukin 12, a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. Interleukin 12 is a disulfide-linked heterodimer composed of the 40 kDa cytokine receptor like subunit encoded by this gene, and a 35 kDa subunit encoded by IL12A. This cytokine is expressed by activated macrophages that serve as an essential inducer of Th1 cells development. This cytokine has been found to be important for sustaining a sufficient number of memory/effector Th1 cells to mediate long-term protection to an intracellular pathogen. Overexpression of this gene was observed in the central nervous system of patients with multiple sclerosis (MS), suggesting a role of this cytokine in the pathogenesis of the disease. The promoter polymorphism of this gene has been reported to be associated with the severity of atopic and non-atopic asthma in children.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CLMF p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL-12 subunit p40; IL-12B; IL12, subunit p40; IL12A; ILN; Interleukin; Interleukin 12; interleukin 12, p40; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 beta chain; Interleukin-12 subunit beta; Interleukin12; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF2; RP23-388G23.1
Gene Aliases: IL12B; NKSF2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support