Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-9523 detects NT4/5 in human and mouse samples.
PA1-9523 has been successfully used in ELISA procedures.
PA1-9523 immunogen is a synthetic peptide based on the human NT4/5 protein.
This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neurotrophic factor 4; neurotrophic factor 5; Neurotrophin 4/5; neurotrophin 5 (neurotrophin 4/5); Neurotrophin-4; Neurotrophin-5; Neutrophic factor 4; Neutrophic factor 5; NT-4; NT-5; NT4/NT5
Gene Aliases: 2900040K06Rik; AI462899; GLC10; GLC1O; NT-4; NT-4/5; NT-5; NT4; NT4/5; NT5; Ntf-5; NTF4; NTF5
UniProt ID: (Human) P34130, (Mouse) Q80VU4
Entrez Gene ID: (Human) 4909, (Mouse) 78405
Molecular Function: neurotrophic factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support