Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
It is not recommended to aliquot this antibody.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B lymphocyte gene 1; E(Sp1) homolog; enhancer of split groucho 3; enhancer of split groucho 4; Enhancer of split groucho-like protein 1; Enhancer of split groucho-like protein 2; Enhancer of split groucho-like protein 3; ESG; ESG-2; ESG-3; ESG-4; ESG1; ESG2; ESG3; Grg-1; Grg-3; Grg-4; groucho-related protein; Groucho-related protein 1; Groucho-related protein 4; transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 2, homolog of Drosophila E(sp1); transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 3 short isoform; transducin-like enhancer of split 3, homolog of Drosophila E(sp1); transducin-like enhancer of split 4 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 4 (homolog of Drosophila E(spl)) - like; transducin-like enhancer of split 4, homolog of Drosophila E(sp1); Transducin-like enhancer protein 1; Transducin-like enhancer protein 2; Transducin-like enhancer protein 3; Transducin-like enhancer protein 4
Gene Aliases: 2610103N05Rik; 5730411M05Rik; AA792082; BCE-1; BCE1; C230057C06Rik; E(spI); ESG; ESG1; ESG2; ESG3; ESG4; ESTM13; ESTM14; Grg-1; Grg-4; GRG1; GRG2; GRG3; Grg3a; Grg3b; GRG4; HsT18976; KIAA1261; KIAA1547; mKIAA1547; mKIAA4188; TLE1; TLE2; TLE3; TLE4; Tle4l; X83332; X83333
UniProt ID: (Human) Q04724, (Human) Q04725, (Human) Q04726, (Human) Q04727, (Mouse) Q62440, (Mouse) Q9WVB2, (Mouse) Q08122, (Mouse) Q62441
Entrez Gene ID: (Human) 7088, (Human) 7089, (Human) 7090, (Human) 7091, (Mouse) 21885, (Mouse) 21886, (Mouse) 21887, (Mouse) 21888
Molecular Function: transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support