Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
This antibody is predicted to react with most mammals, birds, amphibians and fish based on 100% sequence homology.
This antibody contains enough material to conduct 10 mini-Western blots.
Procollagen type III N-terminal peptide (P3NP) is released as part of collagen synthesis process in muscle. The increase of circulating P3NP is a marker not only of muscle growth, but also of muscle repair and fibrosis. P3NP may be a potential biomarker for sarcopenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-1 type 1 collagen; Alpha-1 type I collagen; alpha1(I) procollagen; collagen alpha 1 chain type I; Collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen of skin, tendon and bone, alpha-1 chain; collagen, type 1, alpha 1; collagen, type I, alpha 1; pro-alpha-1 collagen type 1; Procollagen I N; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; type I proalpha 1; type I procollagen alpha 1 chain; type I-alpha 1 collagen
Gene Aliases: Col1a-1; COL1A1; Cola-1; Cola1; COLIA1; EDSC; Mov-13; Mov13; OI1; OI2; OI3; OI4
UniProt ID: (Human) Q15201, (Rat) P02454, (Mouse) P11087
Entrez Gene ID: (Human) 1277, (Rat) 29393, (Mouse) 12842
Molecular Function: extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support