Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLLALENSTYQ YIRVWLERGR LAMLTPNSPK LVVKFVLNDG NVHLISLKIK PYKIELYQSS QNLGFISAST WKIEKGDV
Highest antigen sequence identity to the following orthologs - mouse 75%, rat 68%.
CRB1 (Crumbs homolog 1) plays a role in photoreceptor morphogenesis in the retina. It may maintain cell polarization and adhesion. CRB1 is similar to the Drosophila crumbs protein and localizes to the inner segment of mammalian photoreceptors. The first identified human homolog, CRB1, is expressed in retina and some parts of the brain, leaving room for another homolog to function in epithelial tissues. Mutations in this gene are associated with a severe form of retinitis pigmentosa, RP12, and with Leber congenital amaurosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: crumbs family member 1, photoreceptor morphogenesis associated; Crumbs homolog 1; Protein crumbs homolog 1
Gene Aliases: CRB1; LCA8; RP12
UniProt ID: (Human) P82279
Entrez Gene ID: (Human) 23418
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support