Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Control: A549, Jurkat, NT2D1
Predicted Reactivity: Chimpanzee (100%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; ILN; Interleukin; Interleukin16; lymphocyte chemoattractant factor; M-IL-16; neuronal interleukin 16; Pro-interleukin-16; prointerleukin 16
Gene Aliases: IL16; LCF; NIL16; prIL-16; PRIL16
UniProt ID: (Human) Q14005
Entrez Gene ID: (Human) 3603
Molecular Function: interleukin superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support