Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101378.3 | NP_172962.1 | 1 | 564 | 71 | |
GenBank mRNA | AF078822.1 | - | 1 | 534 | 71 | |
AY075590.1 | - | 1 | 553 | 71 | ||
AY088303.1 | - | 1 | 538 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |