Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCA2;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101426.5 | NP_173010.1 | 8 - 9 | 1356 | 68 | |
GenBank mRNA | AY072105.1 | - | 8 - 9 | 1201 | 68 | |
AY096733.1 | - | 7 - 8 | 1010 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |