Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101603.3 | NP_564021.1 | 6 - 7 | 1923 | 66 | |
GenBank mRNA | AY075625.1 | - | 6 - 7 | 1835 | 66 | |
BT006348.1 | - | 6 - 7 | 1734 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |