Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PnsB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084090.2 | NP_001077559.2 | 1 - 2 | 99 | 93 | |
NM_001123834.1 | NP_001117306.1 | 1 - 2 | 99 | 93 | ||
NM_001123835.4 | NP_001117307.1 | 1 - 2 | 99 | 93 | ||
NM_101731.5 | NP_173308.2 | 1 - 2 | 294 | 93 | ||
GenBank mRNA | AK226496.1 | - | 1 - 2 | 132 | 93 | |
AY080701.1 | - | 1 - 2 | 131 | 93 | ||
AY117165.1 | - | 1 - 2 | 72 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |