Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhca6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101773.3 | NP_173349.1 | 1 - 2 | 457 | 75 | |
GenBank mRNA | AY086461.1 | - | 1 - 2 | 338 | 75 | |
BT020322.1 | - | 1 - 2 | 291 | 75 | ||
BT020554.1 | - | 1 - 2 | 281 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |