Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101780.4 | NP_173356.1 | 7 - 8 | 995 | 75 | |
GenBank mRNA | AB493467.1 | - | 7 - 8 | 641 | 75 | |
AY669794.1 | - | 7 - 8 | 641 | 75 | ||
U53672.2 | - | 7 - 8 | 859 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |