Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WOX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332460.1 | NP_001322716.1 | 1 - 2 | 1183 | 169 | |
NM_101922.3 | NP_173493.2 | 2 - 3 | 597 | 169 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |