Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKB2;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101946.4 | NP_173517.1 | 1 - 2 | 312 | 154 | |
GenBank mRNA | AK229456.1 | - | 1 - 2 | 311 | 154 | |
BT024780.1 | - | 1 - 2 | 230 | 154 |
Set Membership: |
![]() ![]() ![]() ![]() |