Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUC-L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103785.4 | NP_175322.1 | 9 - 10 | 961 | 132 | |
GenBank mRNA | BT005793.1 | - | 9 - 10 | 856 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |