Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126341.3 | NP_178389.1 | 12 - 13 | 1889 | 89 | |
NM_201675.2 | NP_973404.1 | 12 - 13 | 1864 | 89 | ||
GenBank mRNA | AJ289165.1 | - | 12 - 13 | 1566 | 89 | |
AK226970.1 | - | 12 - 13 | 1693 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |