Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111088.4 | NP_186870.1 | 5 - 6 | 1470 | 133 | |
GenBank mRNA | AK118555.1 | - | 5 - 6 | 1140 | 133 | |
BT005373.1 | - | 5 - 6 | 1075 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |